Learn More
Medchemexpress LLC Propargyl-PEG8-NH2 | 1196732-52-1 | 98.0% | 10 G
SDP
Catalog No. 5000340510
Encompass_Preferred
Propargyl-PEG8-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a non-cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Non-cleavable adc linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-40510 Supplier Medchemexpress LLC Supplier No. HY13018210G
Propargyl-PEG8-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a non-cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Non-cleavable adc linker
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)