Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-NH2 | 1196732-52-1 | MFCD28334507 | ≥98.0% | 407.50 | C19H37NO8 | 100 MG
SDP

Catalog No. 5000203829
Encompass_Preferred

Propargyl-PEG8-NH2 is a PEG-based linker reagent used for bioconjugation, including PROTAC and ADC synthesis. It contains a terminal propargyl (alkyne) group and a primary amine, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and amine coupling reactions. The compound is a liquid (colorless to light yellow), with a molecular weight of 407.50 and high purity for research applications.

  • Contains terminal alkyne for CuAAC click chemistry.
  • Contains primary amine for amide and other coupling reactions.
  • PEG8 spacer adds hydrophilicity and flexibility.
  • High purity (≥98.0%).
  • Available in small and bulk package sizes, including 100 mg.
  • Store at 4°C, protect from light, and keep under nitrogen; long-term storage in solvent at -80°C.

Catalog No. 50-002-03829 Supplier Medchemexpress LLC Supplier No. HY130182100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG8-NH2 is a PEG-based linker reagent used for bioconjugation, including PROTAC and ADC synthesis. It contains a terminal propargyl (alkyne) group and a primary amine, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) and amine coupling reactions. The compound is a liquid (colorless to light yellow), with a molecular weight of 407.50 and high purity for research applications.

  • Contains terminal alkyne for CuAAC click chemistry.
  • Contains primary amine for amide and other coupling reactions.
  • PEG8 spacer adds hydrophilicity and flexibility.
  • High purity (≥98.0%).
  • Available in small and bulk package sizes, including 100 mg.
  • Store at 4°C, protect from light, and keep under nitrogen; long-term storage in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.