Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-NH2 | 1196732-52-1 | MFCD28334507 | ≥98.0% | 407.50 g/mol | C19H37NO8 | 1 G
SDP

Catalog No. 5000202605
Encompass_Preferred

Propargyl-PEG8-NH2 is a polyethylene glycol (PEG8) alkyne linker bearing a terminal primary amine. It is used as a non-cleavable linker in conjugation chemistry, including ADC and PROTAC synthesis, and as a click-chemistry building block for copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Alkyne functional group for CuAAC click reactions.
  • Terminal primary amine for conjugation and linker formation.
  • High purity (≥98.0%).
  • Molecular weight 407.50 g/mol.
  • Chemical formula C19H37NO8.
  • Available in multiple package sizes, including 1 G.

Catalog No. 50-002-02605 Supplier Medchemexpress LLC Supplier No. HY1301821G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG8-NH2 is a polyethylene glycol (PEG8) alkyne linker bearing a terminal primary amine. It is used as a non-cleavable linker in conjugation chemistry, including ADC and PROTAC synthesis, and as a click-chemistry building block for copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Alkyne functional group for CuAAC click reactions.
  • Terminal primary amine for conjugation and linker formation.
  • High purity (≥98.0%).
  • Molecular weight 407.50 g/mol.
  • Chemical formula C19H37NO8.
  • Available in multiple package sizes, including 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.