Learn More
Medchemexpress LLC Propargyl-PEG8-NH2 | 1196732-52-1 | MFCD28334507 | >98.0% | 407.50 g/mol | C19H37NO8 | 10 MG
SDP
Catalog No. 5000201991
Encompass_Preferred
Propargyl-PEG8-NH2 is an alkyne-terminated polyethylene glycol (PEG8) amine linker used for bioconjugation and linker synthesis. It provides a terminal alkyne for click chemistry and a primary amine for further derivatization, with a PEG8 spacer to improve solubility and spacing between conjugated moieties.
  • Provides a terminal alkyne group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Contains a primary amine for amide formation and further functionalization.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High chemical purity suitable for research applications.
  • Colorless to light yellow liquid, convenient for solution-phase coupling reactions.
  • Commonly used in PROTAC, ADC, and general bioconjugation workflows.
Catalog No. 50-002-01991 Supplier Medchemexpress LLC Supplier No. HY13018210MG
Propargyl-PEG8-NH2 is an alkyne-terminated polyethylene glycol (PEG8) amine linker used for bioconjugation and linker synthesis. It provides a terminal alkyne for click chemistry and a primary amine for further derivatization, with a PEG8 spacer to improve solubility and spacing between conjugated moieties.
  • Provides a terminal alkyne group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Contains a primary amine for amide formation and further functionalization.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High chemical purity suitable for research applications.
  • Colorless to light yellow liquid, convenient for solution-phase coupling reactions.
  • Commonly used in PROTAC, ADC, and general bioconjugation workflows.