Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-NHS ester | 2182601-74-5 | 95.0% | 533.57 g/mol | C24H39NO12 | 100 MG
SDP

Catalog No. 5000204122
Encompass_Preferred

Propargyl-PEG8-NHS ester is a PEG-based linker and click-chemistry reagent containing a propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is intended for bioconjugation workflows including PROTAC and antibody-drug conjugate linker synthesis.

  • Contains an alkyne (propargyl) group for copper-catalyzed azide-alkyne cycloaddition.
  • Features an NHS ester for efficient coupling to primary amines.
  • Molecular weight 533.57 g/mol and formula C24H39NO12.
  • Typical purity 95.0%.
  • Available in small research quantities (e.g., 100 MG) for bioconjugation studies.
  • Recommended storage: pure form at -20°C to preserve stability.

Catalog No. 50-002-04122 Supplier Medchemexpress LLC Supplier No. HY130376100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG8-NHS ester is a PEG-based linker and click-chemistry reagent containing a propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is intended for bioconjugation workflows including PROTAC and antibody-drug conjugate linker synthesis.

  • Contains an alkyne (propargyl) group for copper-catalyzed azide-alkyne cycloaddition.
  • Features an NHS ester for efficient coupling to primary amines.
  • Molecular weight 533.57 g/mol and formula C24H39NO12.
  • Typical purity 95.0%.
  • Available in small research quantities (e.g., 100 MG) for bioconjugation studies.
  • Recommended storage: pure form at -20°C to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.