Learn More
Medchemexpress LLC Propargyl-PEG8-NHS ester | 2182601-74-5 | 97.0% | 1 G
SDP
Catalog No. 5000339065
Encompass_Preferred
Propargyl-PEG8-NHS ester is a PEG/Alkyl/ether-based PROTAC linker primarily used in the synthesis of PROTACs and as a cleavable ADC linker in the synthesis of antibody-drug conjugates (ADCs). It functions as a click chemistry reagent due to its alkyne group.
  • Contains an alkyne group for chemical reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Suitable for PROTAC synthesis
  • Ideal for antibody-drug conjugate (ADC) synthesis
Catalog No. 50-003-39065 Supplier Medchemexpress LLC Supplier No. HY1303761G
Propargyl-PEG8-NHS ester is a PEG/Alkyl/ether-based PROTAC linker primarily used in the synthesis of PROTACs and as a cleavable ADC linker in the synthesis of antibody-drug conjugates (ADCs). It functions as a click chemistry reagent due to its alkyne group.
  • Contains an alkyne group for chemical reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
  • Suitable for PROTAC synthesis
  • Ideal for antibody-drug conjugate (ADC) synthesis