Learn More
Medchemexpress LLC Propargyl-PEG8-NHS ester | 2182601-74-5 | MFCD31536764 | 95.0% | 533.57 | C24H39NO12 | 50 MG
SDP
Catalog No. 5000204262
Encompass_Preferred
Propargyl-PEG8-NHS ester is an alkyne-terminated PEG8 N-hydroxysuccinimidyl (NHS) ester used as an activated linker for amine coupling and copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Provides an activated NHS ester for efficient amine coupling.
  • Contains a terminal propargyl group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • Suitable for linker and conjugation synthesis such as ADC and PROTAC linker construction.
  • Viscous liquid physical form with recommended cold storage for stability.
Catalog No. 50-002-04262 Supplier Medchemexpress LLC Supplier No. HY13037650MG
Propargyl-PEG8-NHS ester is an alkyne-terminated PEG8 N-hydroxysuccinimidyl (NHS) ester used as an activated linker for amine coupling and copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Provides an activated NHS ester for efficient amine coupling.
  • Contains a terminal propargyl group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • Suitable for linker and conjugation synthesis such as ADC and PROTAC linker construction.
  • Viscous liquid physical form with recommended cold storage for stability.