Learn More
Medchemexpress LLC Propargyl-PEG8-NHS ester | 2182601-74-5 | MFCD31536764 | >95.0% | 533.57 g/mol | C24H39NO12 | 250 MG
SDP
Catalog No. 5000203909
Encompass_Preferred
Propargyl-PEG8-NHS ester is an amine-reactive polyethylene glycol (PEG) linker bearing a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) activated ester. It is used for bioconjugation, click chemistry, and as a linker in ADC/PROTAC synthesis, providing improved solubility and site-selective attachment.
  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Terminal propargyl alkyne enables copper-catalyzed or strain-promoted click reactions.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity typically ≥95% for reliable biochemical use.
  • Molecular weight approximately 533.57 g/mol, suitable for linker applications.
  • Available as a 250 MG solid for laboratory-scale synthesis.
Catalog No. 50-002-03909 Supplier Medchemexpress LLC Supplier No. HY130376250MG
Propargyl-PEG8-NHS ester is an amine-reactive polyethylene glycol (PEG) linker bearing a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) activated ester. It is used for bioconjugation, click chemistry, and as a linker in ADC/PROTAC synthesis, providing improved solubility and site-selective attachment.
  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Terminal propargyl alkyne enables copper-catalyzed or strain-promoted click reactions.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity typically ≥95% for reliable biochemical use.
  • Molecular weight approximately 533.57 g/mol, suitable for linker applications.
  • Available as a 250 MG solid for laboratory-scale synthesis.