Learn More
Medchemexpress LLC Propargyl-PEG8-OH | 1351556-81-4 | 1 G
SDP
Catalog No. 5000338401
Encompass_Preferred
Propargyl-PEG8-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-38401 Supplier Medchemexpress LLC Supplier No. HY1303741G
Propargyl-PEG8-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)