Learn More
Medchemexpress LLC Propargyl-PEG8-OH | 1351556-81-4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130374100MG
Encompass_Preferred
Propargyl-PEG8-OH is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-39230