Learn More
Medchemexpress LLC Propargyl-peg8-oh | 1351556-81-4 | 250 MG
SDP
Catalog No. 5000342002
Encompass_Preferred
Propargyl-PEG8-OH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Utilized in the synthesis of protacs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAC reactions
  • Leverages the intracellular ubiquitin-proteasome system
Catalog No. 50-003-42002 Supplier Medchemexpress LLC Supplier No. 250MGHY130374
Propargyl-PEG8-OH is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Utilized in the synthesis of protacs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAC reactions
  • Leverages the intracellular ubiquitin-proteasome system