Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-OH | 1351556-81-4 | 5 G
SDP

Catalog No. 5000339231
Encompass_Preferred

Propargyl-PEG8-OH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Molecular weight: 408.48
  • Formula: C19H36O9
  • Relevant for cancer and cancer targeted therapy

Catalog No. 50-003-39231 Supplier Medchemexpress LLC Supplier No. HY1303745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-PEG8-OH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Molecular weight: 408.48
  • Formula: C19H36O9
  • Relevant for cancer and cancer targeted therapy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.