Learn More
Medchemexpress LLC Propargyl-PEG9-amine | 2093153-98-9 | 97.0% | 1 G
SDP
Catalog No. 5000339066
Encompass_Preferred
Propargyl-PEG9-amine is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs and functions as a click chemistry reagent, containing an Alkyne group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Supports selective degradation of target proteins
Catalog No. 50-003-39066 Supplier Medchemexpress LLC Supplier No. HY1303731G
Propargyl-PEG9-amine is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs and functions as a click chemistry reagent, containing an Alkyne group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Supports selective degradation of target proteins