Learn More
Medchemexpress LLC Propargyl-PEG9-amine | 2093153-98-9 | 97.0% | 25 MG
SDP
Catalog No. 5000342296
Encompass_Preferred
Propargyl-PEG9-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-42296 Supplier Medchemexpress LLC Supplier No. 25MGHY130373
Propargyl-PEG9-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups