Learn More
Medchemexpress LLC Propargyl-PEG9-amine | 2093153-98-9 | 97.0% | 250 MG
SDP
Catalog No. 5000341492
Encompass_Preferred
Propargyl-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent that contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Click chemistry reagent
  • Alkyne group for reactions
  • Used in PROTAC synthesis
  • Applicable in cancer research
  • Supports cancer targeted therapy
  • PROTAC linker
Catalog No. 50-003-41492 Supplier Medchemexpress LLC Supplier No. 250MGHY130373
Propargyl-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent that contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Click chemistry reagent
  • Alkyne group for reactions
  • Used in PROTAC synthesis
  • Applicable in cancer research
  • Supports cancer targeted therapy
  • PROTAC linker