Learn More
Medchemexpress LLC Propargyl-PEG9-amine | 2093153-98-9 | 97.0% | 50 MG
SDP
Catalog No. 5000339067
Encompass_Preferred
Propargyl-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing Azide groups
Catalog No. 50-003-39067 Supplier Medchemexpress LLC Supplier No. HY13037350MG
Propargyl-PEG9-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules containing Azide groups