Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargylamine

Catalog No. 502907451 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7451 250 g
P509001G 1 g
P5090025G 25 g
P509005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-290-7451 Supplier Sigma Aldrich Supplier No. P50900250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Synthesis of a chiral, fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2450-71-7
Density 0.86 g/mL (at 25°C (literature))
Boiling Point 83°C (lit.)
Molecular Formula C3H5N
Refractive Index n20/D 1.449 (literature)
Linear Formula HC///CCH2NH2
MDL Number MFCD00008198
Quantity 250 g
Synonym 2-Propynylamine; 3-Amino-1-propyne
Molecular Weight (g/mol) 55.08
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.