Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Propargylamine | 250G | 2450-71-7 | MFCD00008198

Catalog No. P50900250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

Propargylamine , 250G

About this item:

CAS #: 2450-71-7
MDL #: MFCD00008198
Purity: 98%
Chemical Formula: C3H5N
Molecular Weight: 55.08
UNSPSC Code: 12352100

Catalog No. P50900250G Supplier Sigma Organic Chemistry Supplier No. P50900250G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargylamine , 250G

About this item:

CAS #: 2450-71-7
MDL #: MFCD00008198
Purity: 98%
Chemical Formula: C3H5N
Molecular Weight: 55.08
UNSPSC Code: 12352100

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.