Learn More
Crescent Chemical Co Inc PROPAZINE 250 MG
SDP
Catalog No. 5076744
Encompass_Preferred
Propazine Herbicide (WS) CAS 139-40-2 Molecular Weight - 229.7098 Storage - RT Catalog Num - DRE-C16440000 pesticide
Catalog No. 50-767-44 Supplier Crescent Chemical Co Inc Supplier No. PSC16440000
Propazine Herbicide (WS) CAS 139-40-2 Molecular Weight - 229.7098 Storage - RT Catalog Num - DRE-C16440000 pesticide