Learn More
Medchemexpress LLC Propazine-d14 | 1219802-87-5 | 99.0% | 1 MG
SDP
Catalog No. 5000412184
Encompass_Preferred
Propazine-d14 is the deuterium labeled Propazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled propazine
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12184 Supplier Medchemexpress LLC Supplier No. HYB1882S1MG
Propazine-d14 is the deuterium labeled Propazine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled propazine
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs