Learn More
Arrow Engineering Type 316 Stainless Steel Propeller 2-1/2"dia. 4 Blade
Catalog No. NC1405904
Encompass_Preferred
The Arrow Mixing Products Type 316 Stainless Steel 2-1/2″ 4 blade propeller with 5/16-24 tapped hole. Our stainless propeller selection is engineered for optimal performance, ensuring smooth and consistent mixing for various industries, including pharmaceuticals, food processing, and chemical production. With a carefully crafted design, these propellers maximize power and efficiency, helping you achieve the best results for your mixing needs.
Catalog No. NC1405904 Supplier Arrow Engineering Supplier No. 25A
The Arrow Mixing Products Type 316 Stainless Steel 2-1/2″ 4 blade propeller with 5/16-24 tapped hole. Our stainless propeller selection is engineered for optimal performance, ensuring smooth and consistent mixing for various industries, including pharmaceuticals, food processing, and chemical production. With a carefully crafted design, these propellers maximize power and efficiency, helping you achieve the best results for your mixing needs.