Learn More
Cayman Chemical PropenylLNIO hydrochloride
SDP
Supplier:  Cayman Chemical 1001172410 MG
Encompass_Preferred
A potent, selective iNOS inhibitor (Kis = 17, 10.3 and 58.2 µM for iNOS, nNOS, and eNOS, respectively, for initial rate binding kinetics); demonstrates reversible tight binding inhibition that is selective for iNOS over nNOS (Kis = 0.56 and 6 µM, respectively)
Catalog No. 50-289-0497