Learn More
Cayman Chemical Propenyl-L-NIO (hydrochloride), Purity: ≥90%, 5 mg
SDP
Catalog No. 502888066
Encompass_Preferred
A potent, selective iNOS inhibitor (Kis = 17, 10.3 and 58.2 µM for iNOS, nNOS, and eNOS, respectively, for initial rate binding kinetics); demonstrates reversible tight binding inhibition that is selective for iNOS over nNOS (Kis = 0.56 and 6 µM, respectively)
Catalog No. 50-288-8066 Supplier Cayman Chemical Supplier No. 100117245 MG
A potent, selective iNOS inhibitor (Kis = 17, 10.3 and 58.2 µM for iNOS, nNOS, and eNOS, respectively, for initial rate binding kinetics); demonstrates reversible tight binding inhibition that is selective for iNOS over nNOS (Kis = 0.56 and 6 µM, respectively)