Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Properdin Factor B Reagent

Catalog No. 22046269 Shop All Beckman Coulter Products
Change view
Click to view available options
No. of Reactions:
150 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
22-046-269 150 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-046-269 Supplier Beckman Coulter™ Supplier No. 447760
Add to Cart
Add to Cart
This item is not returnable. View return policy

Properdin Factor B reagent for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Serum
For Use With (Equipment) Beckman Coulter Analyzers
No. of Reactions 150 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.