Learn More
ACROBiosystems ACROBiosystems Rhesus macaque Properdin Protein, His Tag
Catalog No. 501956404
Encompass_Preferred
Recombinant Protein;100UG;Rhesus macaque Properdin, His Tag (PRN-R52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # F7CGG5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.6 kDa.
Catalog No. 50-195-6404 Supplier ACROBiosystems Supplier No. PRNR52H6100UG
Recombinant Protein;100UG;Rhesus macaque Properdin, His Tag (PRN-R52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # F7CGG5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.6 kDa.