Learn More
ACROBiosystems ACROBiosystems Mouse Properdin Protein, His Tag
Catalog No. 501956406
Encompass_Preferred
Recombinant Protein;100UG;Mouse Properdin, His Tag (PRN-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Pro 464 (Accession # P11680-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.8 kDa. The protein migrates as 58-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6406 Supplier ACROBiosystems Supplier No. PRNM52H5100UG
Recombinant Protein;100UG;Mouse Properdin, His Tag (PRN-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Pro 464 (Accession # P11680-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.8 kDa. The protein migrates as 58-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.