Learn More
ACROBiosystems ACROBiosystems Human Properdin Protein, His Tag
Catalog No. 501957008
Encompass_Preferred
Recombinant Protein;1MG;Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # P27918-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.7 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7008 Supplier ACROBiosystems Supplier No. PRNH52H31MG
Recombinant Protein;1MG;Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # P27918-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.7 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.