Learn More
ACROBiosystems ACROBiosystems Human Properdin Protein, His Tag
Supplier:  ACROBiosystems PRNH52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession # P27918-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.7 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7009