Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Properties of Antacids
GREENER_CHOICESCI_ED

Catalog No. S96651 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96651 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, and Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S96651 Supplier Innovating Science™ Supplier No. IS9352
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Acetyl Salicylic Acid/ Aspirin, and Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Includes

  • 30mL Hydrochloric Acid solution, 1M UN1789 (5)
  • 30mL universal indicator UN1170 (2)
  • 30mL Bromothymol Blue, 0.5% aqueous solution (2)
  • Antacid #1 (7 tablets)
  • Antacid #2 (7 extra-strength)
  • Antacid #3 (7 tablets)
  • Antacid #4 (14 tablets)
  • 60mL Antacid #5
  • Graduated plastic pipets (15)
  • Universal indicator strips (100)
  • Disposable medicine cups (90)
  • Teacher's manual and student study guide copy-masters

Requires

Lab timers (or stopwatches with second hand), 10mL graduated cylinders, Distilled water, Glass rods (or wooden stirring sticks), Electronic balances (optional).

Investigate the more common active components in over-the-counter antacids.

  • Investigate the rate of acid neutralization of five different antacids
  • Compare the buffering ability of three different calcium carbonate-based antacids
  • Enough materials for 15 tests

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To investigate the more common active components in over the counter antacids
Includes 5 x 30mL Hydrochloric Acid, 2 x 30mL universal indicator, 2 x 30mL Bromothymol blue, 0.5% Aqueous Solution, 1 x 60mL #5 Antacid, liquid, #1 Antacid, Tablet, #2 Antacid, Extra strength, #3 Antacid, tablet, #4 Antacid, tablets, graduated Plastic pipettes, u
Quantity 1
Dimensions (L x W x H) 12 x 9.25 x 4 in.
Class Size 15 Lab Groups
Product Type Properties of Antacids Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.