Learn More
Alkali Scientific ProPette™ LE 12-Channel Pipette, 2 to 20µl
SDP
Catalog No. 502336075
Encompass_Preferred
Ideal for all 96 well applications, the new ProPette™ LE 12-Channel Pipette is light weight, rugged, and stands up to the demands of everyday use in a busy laboratory environment. 2 to 20µL usable range. 1/pk
Catalog No. 50-233-6075 Supplier Alkali Scientific Supplier No. P951220U
Ideal for all 96 well applications, the new ProPette™ LE 12-Channel Pipette is light weight, rugged, and stands up to the demands of everyday use in a busy laboratory environment. 2 to 20µL usable range. 1/pk