Learn More
Alkali Scientific ProPette™ Pipette Controller with QuickStand™, Euro Two Prong Plug
SDP
Catalog No. 502335900
Encompass_Preferred
ProPette™ Controllers are lightweight (only 6.3oz/170g) and designed to fit comfortably in either the right or left hand. The natural feel of the ergonomic body design, the computer balanced weight distribution and the low pressure fingertip control buttons all combine to provide a more comfortable pipetting experience. And now the ProPette™ controller includes the patent-pending Quickstand retractable stand. Which allows the instrument to be set down on the bench without risk of contamination. The ProPette™ Controller has variable speed control, is cordless and rechargable, has eight hours of continuous operation, and works with pipettes from 1mL to 100mL. One per pack.
Catalog No. 50-233-5900 Supplier Alkali Scientific Supplier No. P70801
ProPette™ Controllers are lightweight (only 6.3oz/170g) and designed to fit comfortably in either the right or left hand. The natural feel of the ergonomic body design, the computer balanced weight distribution and the low pressure fingertip control buttons all combine to provide a more comfortable pipetting experience. And now the ProPette™ controller includes the patent-pending Quickstand retractable stand. Which allows the instrument to be set down on the bench without risk of contamination. The ProPette™ Controller has variable speed control, is cordless and rechargable, has eight hours of continuous operation, and works with pipettes from 1mL to 100mL. One per pack.