Learn More
MTC Bio ProPette™ Pipette Controller, w/ Digital Controls, w/ QuickStand, USB-C Plug
Supplier:  MTC Bio P6290
Encompass_Preferred
ProPette™ Controllers from MTC Bio™ are lightweight (only 6.3oz/170g) and designed to fit comfortably in either the right or left hand. The natural feel of the ergonomic body design, the computer balanced weight distribution and the low pressure fingertip control button all combine to provide a more comfortable pipetting experience. The New ProPette Digital features enhanced digital controls that allow users to set their maximum aspirate and dispense rates. Simply tap the button to your desired speed on the digital display and a full press of the trigger will match. It also features the popular QuickStand™ retractable stand, which allows the instrument to be set down on the bench without risk of contamination. • With QuickStand™ retractable stand• Variable speed control with digital rate limiter• Cordless and rechargeable• 8 hours of continuous operation• For pipettes from 1mL to 100mL
Catalog No. NC3737136