Learn More
Electron Microscopy Sciences ProPette™ Pipette Controller European Plug
SDP
Catalog No. 501929265
Encompass_Preferred
ProPette Controller's ergonomic fit in the left or right hand, adjustable speed and long battery life, make it the logical choice for comfort, convenience and value. The natural feel of the ergonomic body design, the computer balanced weight distribution, and the low pressure fingertip control buttons, all combine to provide a more comfortable pipetting experience. A rechargable lithium battery keeps the controller running for up to 8 hours or 2500 pipettings on a single two hour charge. ProPette's easy-fit silicone nosepiece accepts pippettes from 1 ml to 100 ml. Includes charger, benchtop stand, extra filter, and replaceable lithium battery.
Catalog No. 50-192-9265 Supplier Electron Microscopy Sciences Supplier No. 7097802
ProPette Controller's ergonomic fit in the left or right hand, adjustable speed and long battery life, make it the logical choice for comfort, convenience and value. The natural feel of the ergonomic body design, the computer balanced weight distribution, and the low pressure fingertip control buttons, all combine to provide a more comfortable pipetting experience. A rechargable lithium battery keeps the controller running for up to 8 hours or 2500 pipettings on a single two hour charge. ProPette's easy-fit silicone nosepiece accepts pippettes from 1 ml to 100 ml. Includes charger, benchtop stand, extra filter, and replaceable lithium battery.