Learn More
Med Vet International Prophy Paste Xylitol Free, J0452D5Q, 200/Box
Catalog No. 5000584655
Encompass_Preferred
Prophy Paste Xylitol Free. Most human prophy paste contain Xylitol which is a known toxic compound to dogs. JorVet prophy paste is Xylitol free! 200 cups/box.
Catalog No. 50-005-84655 Supplier Med Vet International Supplier No. JOR452D5Q
Prophy Paste Xylitol Free. Most human prophy paste contain Xylitol which is a known toxic compound to dogs. JorVet prophy paste is Xylitol free! 200 cups/box.