Learn More
Medchemexpress LLC Propiconazole | 60207-90-1 | 98.9% | 250 MG
SDP
Catalog No. 5000405736
Encompass_Preferred
Propiconazole is an orally active N-substituted triazole used as a fungicide. It has been identified as a mouse liver hepatotoxicant and a hepatocarcinogen, exhibiting adverse reproductive and developmental toxicities in experimental animals.
  • Orally active N-substituted triazole fungicide
  • Identified as a mouse liver hepatotoxicant
  • Exhibits adverse reproductive and developmental toxicities
  • Can induce specific hepatic P450 isoforms
Catalog No. 50-004-05736 Supplier Medchemexpress LLC Supplier No. 250MGHYB0847
Propiconazole is an orally active N-substituted triazole used as a fungicide. It has been identified as a mouse liver hepatotoxicant and a hepatocarcinogen, exhibiting adverse reproductive and developmental toxicities in experimental animals.
  • Orally active N-substituted triazole fungicide
  • Identified as a mouse liver hepatotoxicant
  • Exhibits adverse reproductive and developmental toxicities
  • Can induce specific hepatic P450 isoforms