Learn More
Medchemexpress LLC Propiconazole | 60207-90-1 | 98.91% | 100 MG
SDP
Catalog No. 5000408983
Encompass_Preferred
Propiconazole is an orally active N-substituted triazole used as a fungicide. It is a mouse liver hepatotoxicant and a hepatocarcinogen that has adverse reproductive and developmental toxicities in experimental animals. This product is for research use only and is not sold to patients.
  • Orally active N-substituted triazole
  • Fungicide
  • Mouse liver hepatotoxicant and hepatocarcinogen in experimental animals
  • Associated with adverse reproductive and developmental toxicities in experimental animals
Catalog No. 50-004-08983 Supplier Medchemexpress LLC Supplier No. HYB0847100MG
Propiconazole is an orally active N-substituted triazole used as a fungicide. It is a mouse liver hepatotoxicant and a hepatocarcinogen that has adverse reproductive and developmental toxicities in experimental animals. This product is for research use only and is not sold to patients.
  • Orally active N-substituted triazole
  • Fungicide
  • Mouse liver hepatotoxicant and hepatocarcinogen in experimental animals
  • Associated with adverse reproductive and developmental toxicities in experimental animals