Learn More
AnaSpec Propidium iodide, 1.0 mg/mL solution in water, | MW 668.4 | (10 mL)
Catalog No. 50850556
Encompass
Propidium iodide, 1.0 mg/mL solution in water, ; MW 668.4; (10 mL) Used as a nuclear or chromosome counterstain; stain for dead cells
Catalog No. 50-850-556 Supplier AnaSpec Supplier No. AS83215
Propidium iodide, 1.0 mg/mL solution in water, ; MW 668.4; (10 mL) Used as a nuclear or chromosome counterstain; stain for dead cells