Learn More
Cayman Chemical Propidium Iodide 100MG
SDP
Catalog No. 502955348
Encompass_Preferred
A fluorescent probe commonly used to identify dead cells; excitation at 488-535 nm and emission maximum of 617 nm
Catalog No. 50-295-5348 Supplier Cayman Chemical Supplier No. 14289100 MG
A fluorescent probe commonly used to identify dead cells; excitation at 488-535 nm and emission maximum of 617 nm