Learn More
Cayman Chemical Propidium Iodide, Purity: ≥90%, CAS: 25535-16-4, 10 mg
SDP
Catalog No. 502954443
Encompass_Preferred
A fluorescent probe commonly used to identify dead cells; excitation at 488-535 nm and emission maximum of 617 nm
Catalog No. 50-295-4443 Supplier Cayman Chemical Supplier No. 1428910 MG
A fluorescent probe commonly used to identify dead cells; excitation at 488-535 nm and emission maximum of 617 nm