Learn More
Medchemexpress LLC HY-D0815 10mg Medchemexpress, Propidium (Iodide) CAS:25535-16-4 Purity:>98%
SDP
Catalog No. 501933184
Encompass_Preferred
Medchemexpress, HY-D0815 10mg Propidium (Iodide) CAS:25535-16-4 Propidium Iodide is a red-fluorescent dye that can be used to stain cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3184 Supplier Medchemexpress LLC Supplier No. HYD081510MG
Medchemexpress, HY-D0815 10mg Propidium (Iodide) CAS:25535-16-4 Propidium Iodide is a red-fluorescent dye that can be used to stain cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.