Learn More
AdipoGen Propidium iodide, 25535-16-4, MFCD00011921, 25 mg
SDP
Catalog No. 50596071
Encompass_Preferred
MF: C27H34I2N4, MW: 668.39, ≥94%, Appearance: Red powder. Propidium iodide is the most common red-fluorescent nuclear stain. It can also be excited using excitation filters appropriate for green-fluorescent dyes.
Catalog No. 50-596-071 Supplier AdipoGen Supplier No. CDXP0023M025
MF: C27H34I2N4, MW: 668.39, ≥94%, Appearance: Red powder. Propidium iodide is the most common red-fluorescent nuclear stain. It can also be excited using excitation filters appropriate for green-fluorescent dyes.