Learn More
Medchemexpress LLC Propidium Iodide | 25535-16-4 | 99.69% | 100 MG
SDP
Catalog No. 5000410926
Encompass_Preferred
Propidium Iodide (PI) is a nuclear staining agent that stains DNA. It is an analogue of ethidine bromide that emits red fluorescence when embedded in double-stranded DNA. PI cannot pass through living cell membranes, but it can pass through damaged cell membranes to stain the nucleus. It has a fluorescence wavelength of 493/617 nm and 536/635 nm after Mosaic with DNA. Propidium Iodide is commonly used in the detection of apoptosis or necrosis, and is often used in flow cytometry analysis.
  • Nuclear staining agent for DNA.
  • Emits red fluorescence upon embedding in double-stranded DNA.
  • Passes through damaged cell membranes to stain the nucleus.
  • Fluorescence wavelength: 493/617 nm and 536/635 nm (after Mosaic with DNA).
  • Commonly used in the detection of apoptosis or necrosis.
  • Often used in flow cytometry analysis.
  • Used in cell cycle and cell viability assays.
Catalog No. 50-004-10926 Supplier Medchemexpress LLC Supplier No. HYD0815100MG
Propidium Iodide (PI) is a nuclear staining agent that stains DNA. It is an analogue of ethidine bromide that emits red fluorescence when embedded in double-stranded DNA. PI cannot pass through living cell membranes, but it can pass through damaged cell membranes to stain the nucleus. It has a fluorescence wavelength of 493/617 nm and 536/635 nm after Mosaic with DNA. Propidium Iodide is commonly used in the detection of apoptosis or necrosis, and is often used in flow cytometry analysis.
  • Nuclear staining agent for DNA.
  • Emits red fluorescence upon embedding in double-stranded DNA.
  • Passes through damaged cell membranes to stain the nucleus.
  • Fluorescence wavelength: 493/617 nm and 536/635 nm (after Mosaic with DNA).
  • Commonly used in the detection of apoptosis or necrosis.
  • Often used in flow cytometry analysis.
  • Used in cell cycle and cell viability assays.