Learn More
Medchemexpress LLC Propidium Iodide | 25535-16-4 | 99.7% | 50 MG
SDP
Catalog No. 5000409832
Encompass_Preferred
Propidium Iodide (PI) is a nuclear staining agent that stains DNA. It is an analogue of ethidine bromide, which emits red fluorescence upon embedding in double-stranded DNA. Propidium Iodide cannot pass through living cell membranes, but it can pass through damaged cell membranes to stain the nucleus. It is commonly used in the detection of apoptosis or necrosis, and is often used in flow cytometry analysis.
  • Nuclear staining agent for DNA.
  • Emits red fluorescence upon embedding in double-stranded DNA.
  • Cannot pass through living cell membranes, but stains nuclei of cells with damaged membranes.
  • Fluorescence wavelength of 493/617 nm, and 536/635 nm after Mosaic with DNA.
  • Commonly used in detection of apoptosis or necrosis.
  • Often used in flow cytometry analysis.
  • Can be used in cell cycle and cell viability assays.
Catalog No. 50-004-09832 Supplier Medchemexpress LLC Supplier No. HYD081550MG
Propidium Iodide (PI) is a nuclear staining agent that stains DNA. It is an analogue of ethidine bromide, which emits red fluorescence upon embedding in double-stranded DNA. Propidium Iodide cannot pass through living cell membranes, but it can pass through damaged cell membranes to stain the nucleus. It is commonly used in the detection of apoptosis or necrosis, and is often used in flow cytometry analysis.
  • Nuclear staining agent for DNA.
  • Emits red fluorescence upon embedding in double-stranded DNA.
  • Cannot pass through living cell membranes, but stains nuclei of cells with damaged membranes.
  • Fluorescence wavelength of 493/617 nm, and 536/635 nm after Mosaic with DNA.
  • Commonly used in detection of apoptosis or necrosis.
  • Often used in flow cytometry analysis.
  • Can be used in cell cycle and cell viability assays.