Learn More
Cayman Chemical PropIdIum IodIde 50mg
SDP
Catalog No. 502954560
Encompass_Preferred
A fluorescent probe commonly used to identify dead cells; excitation at 488-535 nm and emission maximum of 617 nm
Catalog No. 50-295-4560 Supplier Cayman Chemical Supplier No. 1428950 MG
A fluorescent probe commonly used to identify dead cells; excitation at 488-535 nm and emission maximum of 617 nm