Learn More
AdipoGen Propidium iodide
SDP
Catalog No. 502560271
Encompass_Preferred
Chemical. CAS 25535-16-4. Formula C27H34I2N4. MW 668.39. Synthetic. Propidium iodide is the most common red-fluorescent nuclear stain. It can also be excited using excitation filters appropriate for green-fluorescent dyes.
Catalog No. 50-256-0271 Supplier AdipoGen Supplier No. CDXP0023G005
Chemical. CAS 25535-16-4. Formula C27H34I2N4. MW 668.39. Synthetic. Propidium iodide is the most common red-fluorescent nuclear stain. It can also be excited using excitation filters appropriate for green-fluorescent dyes.