Learn More
Selleck Chemical LLC Propidium Iodide
SDP
Catalog No. 5000002584
Encompass_Preferred
Propidium iodide (PI) is a red-fluorescent DNA stain which penetrates only damaged cellular membranes Propidium iodide (PI) is described for use in the determination of cell viability in cell suspension
Catalog No. 50-000-02584 Supplier Selleck Chemical LLC Supplier No. S687425MG
Propidium iodide (PI) is a red-fluorescent DNA stain which penetrates only damaged cellular membranes Propidium iodide (PI) is described for use in the determination of cell viability in cell suspension