Learn More
Selleck Chemical LLC Propidium Iodide
Supplier: Selleck Chemical LLC S68745MG
Propidium iodide (PI) is a red-fluorescent DNA stain which penetrates only damaged cellular membranes Propidium iodide (PI) is described for use in the determination of cell viability in cell suspension