Learn More
Crescent Chemical Co Inc PROPIDIUM IODIDE RESEARCH GRDE
SDP
Catalog No. 50248061
Encompass_Preferred
Propidium iodide research grade For selective labelling of DNA in dead cells; used in tumor diagnosis (flow cytometry) (1, 2). For the study of micronuclear morphology of protozoa (3). Size - 25MG Storage Conditions - +2 °C TO +8 °C Catalog Number -
Catalog No. 50-248-061 Supplier Crescent Chemical Co Inc Supplier No. SE33671.01
Propidium iodide research grade For selective labelling of DNA in dead cells; used in tumor diagnosis (flow cytometry) (1, 2). For the study of micronuclear morphology of protozoa (3). Size - 25MG Storage Conditions - +2 °C TO +8 °C Catalog Number -