Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propidium Iodide Solution, Sigma-Aldrich

Catalog No. 01258886 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-258-886 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-886 Supplier MilliporeSigma Supelco Supplier No. P486410ML
Add to Cart
Add to Cart

2024 CiteAb Award Winner for Supplier Succeeding in Parkinson's Research

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 mL
Form Liquid
Concentration ≥94%
CAS 25535-16-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.