Learn More
BIOGEMS INTERNATIONAL INC Propidium Iodide Solution 200 tests
SDP
Catalog No. 501530205
Encompass
The PI (Propidium iodide) solution can be used to exclude non-viable cells in flow cytometry assays. PI is a fluorescent dye that binds to double stranded DNA in cells that have lost plasma membrane integrity, such as those in the late stages of apoptosis or dead cells. It is detected in the orange 562-588 nm band-pass filter.
Catalog No. 50-153-0205 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 6091000200
Add to Cart
Add to Cart
The PI (Propidium iodide) solution can be used to exclude non-viable cells in flow cytometry assays. PI is a fluorescent dye that binds to double stranded DNA in cells that have lost plasma membrane integrity, such as those in the late stages of apoptosis or dead cells. It is detected in the orange 562-588 nm band-pass filter.